CAS RN 156339-88-7
CAS RN 156339-88-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Albuterol Aldehyde Hemisulfate, TRC
CAS: 156339-88-7 Molecular Formula: C13 H19 N O3 Molecular Weight (g/mol): 237.29 Synonym: 5-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-2-hydroxybenzaldehyde,Salbutamol Sulphate Imp. D (EP) IUPAC Name: 5-[2-(tert-butylamino)-1-hydroxyethyl]-2-hydroxybenzaldehyde SMILES: CC(C)(C)NCC(O)c1ccc(O)c(C=O)c1