accessibility menu, dialog, popup

UserName

CH3HOO(E)

CAS RN 1577-18-0

IUPAC Name: (3E)-hex-3-enoic acid
Synonyms: trans-3-hexenoic acid hex-3t-enoic acid hydrosorbic acid trans-3-hexanoic acid trans-hex-3-ensaeure
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: XXHDAWYDNSXJQM-ONEGZZNKSA-N
SMILES: CC\C=C\CC(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results