Learn More
trans-3-Hexenoic acid, 97%
Product Code. 11445473
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445473 50 g 50g
11435473 10 g 10g
11455473 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11445473 Supplier Thermo Scientific Alfa Aesar Supplier No. A13428.18
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 1577-18-0 | C6H10O2 | 114.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1577-18-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00002786
InChI Key XXHDAWYDNSXJQM-ONEGZZNKSA-N
Synonym trans-3-hexenoic acid, hydrosorbic acid, 3-hexenoic acid, e-hex-3-enoic acid, 3-hexenoic acid, 3e, e-3-hexenoic acid, 3-hexenoic acid, e, trans-hex-3-enoic acid, unii-9b3n069edg, fema no. 3170
PubChem CID 5282708
ChEBI CHEBI:49285
IUPAC Name (E)-hex-3-enoic acid
SMILES CC\C=C\CC(O)=O
Specifications
Melting Point 11°C to 12°C
Density 0.961
Boiling Point 206°C
Flash Point 99°C (210°F)
Odor Rancid
Refractive Index 1.4398
Quantity 50 g
UN Number UN3265
Beilstein 1700862
Formula Weight 114.14
Percent Purity 97%
Chemical Name or Material trans-3-Hexenoic acid
Show More Show Less
compliance-icons
Product Identifier
  • trans-3-Hexenoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: trans-3-Hexenoic acid