accessibility menu, dialog, popup

UserName

ONON

CAS RN 15822-77-2

IUPAC Name: 1-(2-nitrophenyl)piperidine
Synonyms: 1-(2-nitrophenyl)piperidine
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: WDAUAKBDZSYXEM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results