missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Nitrophenyl)piperidine, 98%, Thermo Scientific™

Product Code. 11496147
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496147 25 g 25g
11486147 5 g 5g
11406157 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496147 Supplier Thermo Scientific Alfa Aesar Supplier No. B25116.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15822-77-2
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD00023653
InChI Key WDAUAKBDZSYXEM-UHFFFAOYSA-N
Synonym 1-2-nitrophenyl piperidine, 2-nitrophenyl piperidine, pubchem10243, maybridge1_000639, acmc-1c4am, cbmicro_018648, cambridge id 5345369, 1-o-nitrophenyl piperidine, 1-2-nitrophenyl piperidine #, piperidine,1-2-nitrophenyl
PubChem CID 225457
IUPAC Name 1-(2-nitrophenyl)piperidine
SMILES C1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]

Specifications

Melting Point 76°C to 78°C
Quantity 25 g
Formula Weight 206.24
Percent Purity 98%
Chemical Name or Material 1-(2-Nitrophenyl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.