CAS RN 15894-70-9
CAS RN 15894-70-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1,1'-(Hexane-1,6-diyl)bis[3-cyanoguanidine], Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
1,6-Bis(cyano-guanidino)hexane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,6-Bis(cyano-guanidino)hexane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.