accessibility menu, dialog, popup

UserName

CAS RN 1604-28-0

IUPAC Name: (3E)-6-methylhepta-3,5-dien-2-one
Synonyms: methylheptadienone methyl heptadienone
Molecular Weight (g/mol): 124.18
Molecular Formula: C8H12O
InChi Key: KSKXSFZGARKWOW-GQCTYLIASA-N
SMILES: CC(C)=C\C=C\C(C)=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results