CAS RN 1610882-30-8
CAS RN 1610882-30-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ TCS OX2 29
Tocris Bioscience™ TCS OX2 29
CAS: 1610882-30-8 Molecular Formula: C23H32ClN3O3