CAS RN 1627696-51-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
LY3177833, MedChemExpress
MedChemExpress LY3177833 is a CDC7 and pMCM2 inhibitor with IC50 values of 3.3 nM and 290 nM, respectively.