accessibility menu, dialog, popup

UserName

CAS RN 16314-18-4

CH3CH3CH3OHC

CAS RN 16314-18-4

IUPAC Name: 3-(tert-butoxy)prop-1-yne
Synonyms: 3-(tert-butoxy)prop-1-yne
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: QIGSPXPODKLGRK-UHFFFAOYSA-N
SMILES: CC(C)(C)OCC#C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)