CAS RN 1637739-82-2
CAS RN 1637739-82-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
BQU57, MedChemExpress
MedChemExpress BQU57 shows selective inhibition for Ral relative to Ras or Rho and inhibit xenograft tumor growth similar to depletion of Ral by siRNA. The IC50 for BQU57 of 2.0 (mu)M in H2122 and 1.3 (mu)M in H358.