accessibility menu, dialog, popup

UserName

CAS RN 16727-44-9

IUPAC Name: 3,5-dibromo-2,6-dimethoxypyridine
Synonyms: 3,5-dibromo-2,6-dimethoxypyridine
Molecular Weight (g/mol): 296.95
Molecular Formula: C7H7Br2NO2
InChi Key: SQWUBNSHUMRETN-UHFFFAOYSA-N
SMILES: COC1=NC(OC)=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results