missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-2,6-dimethoxypyridine, 99%, Thermo Scientific™

Product Code. 11496510
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496510 25 g 25g
11486510 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496510 Supplier Thermo Scientific Alfa Aesar Supplier No. H27598.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16727-44-9
Molecular Formula C7H7Br2NO2
Molecular Weight (g/mol) 296.946
MDL Number MFCD00955954
InChI Key SQWUBNSHUMRETN-UHFFFAOYSA-N
Synonym pyridine,3,5-dibromo-2,6-dimethoxy
PubChem CID 42553050
IUPAC Name 3,5-dibromo-2,6-dimethoxypyridine
SMILES COC1=C(C=C(C(=N1)OC)Br)Br

Specifications

Melting Point 85°C to 87°C
Quantity 25 g
Formula Weight 296.95
Percent Purity 99%
Chemical Name or Material 3,5-Dibromo-2,6-dimethoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.