accessibility menu, dialog, popup

UserName

CAS RN 16806-93-2

OO

CAS RN 16806-93-2

IUPAC Name: 4,5,6,7-tetrahydro-1-benzofuran-4-one
Synonyms: 6,7-dihydro-5H-1-benzofuran-4-one
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: DXWQOYPYNPSVRL-UHFFFAOYSA-N
SMILES: O=C1CCCC2=C1C=CO2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results