accessibility menu, dialog, popup

UserName

FFFFSClOO

CAS RN 1682-10-6

IUPAC Name: 4-fluoro-3-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 4-fluoro-3-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 262.6
Molecular Formula: C7H3ClF4O2S
InChi Key: AYOKQWBPSUHEQU-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1C(F)(F)F)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results