missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Artikelnummer. 15457575
Ansicht ändern
Click to view available options
Quantity:
250 mg
1 g
Packungsgröße:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
15457575 250 mg 250mg
11421300 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Product Code. 15457575 Supplier Thermo Scientific Alfa Aesar Supplier No. H26372.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1682-10-6
Molecular Formula C7H3ClF4O2S
Molecular Weight (g/mol) 262.603
MDL Number MFCD01091001
InChI Key AYOKQWBPSUHEQU-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl benzenesulfonyl chloride, 4-fluoro-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-fluoro-3-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro-3-trifluoromethyl, 5-chlorosulphonyl-2-fluorobenzotrifluoride, 4-fluoro-3-trifluoromethyl benzenesulfonylchloride, 4-fluoro-3-trifluoromethyl-benzenesulfonyl chloride, pubchem8507, acmc-209vib, ksc171m7b
PubChem CID 2737549
IUPAC Name 4-fluoro-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)F

Specifications

Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 262.61
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.