accessibility menu, dialog, popup

UserName

CAS RN 16903-35-8

NH4ClClClClAu

CAS RN 16903-35-8

IUPAC Name: gold(3+) ammonium tetrachloride
Synonyms: chloroauric acid
Molecular Weight (g/mol): 356.81
Molecular Formula: AuCl4H4N
InChi Key: WPEJSSRSFRWYJB-UHFFFAOYSA-K
SMILES: [NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Au+3]
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results