accessibility menu, dialog, popup

UserName

CAS RN 16924-03-1

FFFFFFFKKNb

CAS RN 16924-03-1

IUPAC Name: niobium(5+) dipotassium heptafluoride
Synonyms: niobium(5+) dipotassium heptafluoride
Molecular Weight (g/mol): 304.09
Molecular Formula: F7K2Nb
InChi Key: FTPUNAWAGWERLA-UHFFFAOYSA-G
SMILES: [F-].[F-].[F-].[F-].[F-].[F-].[F-].[K+].[K+].[Nb+5]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results