CAS RN 1707147-26-9
CAS RN 1707147-26-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress THK5351
MedChemExpress THK5351 can be radiolabeled and used as a radiotracer for in vivo imaging of tau pathology in the brain.