CAS RN 17078-27-2
IUPAC Name:
bis[4-(dimethylamino)phenyl]ethane-1,2-dione
Synonyms:
bis[4-(dimethylamino)phenyl]ethane-1,2-dione
Molecular Weight (g/mol):
296.37
Molecular Formula:
C18H20N2O2
InChi Key:
AVFUVYIDYFXFSX-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(C=C1)N(C)C
1
–
1
of
1
results
1
4,4'-Bis(dimethylamino)benzil, 98%
CAS: 17078-27-2 Molecular Formula: C18H20N2O2 Molecular Weight (g/mol): 296.36 InChI Key: AVFUVYIDYFXFSX-UHFFFAOYSA-N Synonym: 4,4'-bis dimethylamino benzil,1,2-bis 4-dimethylamino phenyl ethane-1,2-dione,ethandione, bis 4-dimethylamino phenyl,bis 4-dimethylamino phenyl ethane-1,2-dione,acmc-209e1j,benzil-based compound, 35,bis n,n-dimethylamino benzil,4,4/'-bis dimethylamino benzil,bis 4-dimethylamino phenyl diketone PubChem CID: 222570 IUPAC Name: 1,2-bis[4-(dimethylamino)phenyl]ethane-1,2-dione SMILES: CN(C)C1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)N(C)C