accessibility menu, dialog, popup

UserName

CH3NNHSOOCH3(E)

CAS RN 17336-66-2

IUPAC Name: 4-methyl-N'-[(1E)-1-phenylpropylidene]benzene-1-sulfonohydrazide
Synonyms: 4-methyl-N'-[(1E)-1-phenylpropylidene]benzenesulfonohydrazide
Molecular Weight (g/mol): 302.39
Molecular Formula: C16H18N2O2S
InChi Key: TWTWWQDWKPRWFI-WUKNDPDISA-N
SMILES: CC\C(=N/NS(=O)(=O)C1=CC=C(C)C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results