accessibility menu, dialog, popup

UserName

CAS RN 175203-97-1

H3CSClOBr

CAS RN 175203-97-1

IUPAC Name: 2-bromo-1-(5-chloro-3-methyl-1-benzothiophen-2-yl)ethan-1-one
Synonyms: 2-bromo-1-(5-chloro-3-methyl-1-benzothiophen-2-yl)ethanone
Molecular Weight (g/mol): 303.6
Molecular Formula: C11H8BrClOS
InChi Key: HBNULIMIUQEVFS-UHFFFAOYSA-N
SMILES: CC1=C(SC2=CC=C(Cl)C=C12)C(=O)CBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results