accessibility menu, dialog, popup

UserName

CAS RN 1768-64-5

ClO

CAS RN 1768-64-5

IUPAC Name: 4-chlorooxane
Synonyms: 4-chlorooxane
Molecular Weight (g/mol): 120.58
Molecular Formula: C5H9ClO
InChi Key: DHRSKOBIDIDMJZ-UHFFFAOYSA-N
SMILES: ClC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results