accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 17695-48-6

IUPAC Name: 4-methyl-2-oxo-2H-chromen-7-yl hexadecanoate
Synonyms: 4-methyl-2-oxochromen-7-yl hexadecanoate
Molecular Weight (g/mol): 414.59
Molecular Formula: C26H38O4
InChi Key: QKQVFFACWLFRRX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC1=CC=C2C(C)=CC(=O)OC2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results