missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl Palmitate, TRC

Product Code. 30473003
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473003 2.5 g 2.5g
30464835 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30473003 Supplier Toronto Research Chemicals Supplier No. TRCM3330702.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17695-48-6
Molecular Formula C26 H38 O4
Molecular Weight (g/mol) 414.58
Synonym Hexadecanoic Acid 4-Methyl-2-oxo-2H-1-benzopyran-7-yl Ester, 7-Hydroxy-4-methylcoumarin Palmitate, 4-Methylumbelliferone Palmitate, Palmitoyl 4-Methylumbelliferone
IUPAC Name (4-methyl-2-oxochromen-7-yl) hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)Oc1ccc2C(=CC(=O)Oc2c1)C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C26H38O4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-25(27)29-22-17-18-23-21(2)19-26(28)30-24(23)20-22/h17-20H,3-16H2,1-2H3
Formula Weight 414.277
Chemical Name or Material 4-Methylumbelliferyl Palmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.