CAS RN 1794940-24-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methyl Buphedrone-d3 Hydrochloride, TRC
CAS: 1794940-24-1 Molecular Formula: C12H15D3ClNO Molecular Weight (g/mol): 230.75 Synonym: 2-[(Methyl-d3)amino]-1-(4-methylphenyl)-1-butanone Hydrochloride IUPAC Name: 1-(4-methylphenyl)-2-(trideuteriomethylamino)butan-1-one;hydrochloride SMILES: O=C(C(NC([2H])([2H])[2H])CC)C1=CC=C(C)C=C1.Cl