CAS RN 1797134-47-4
CAS RN 1797134-47-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-(tert-Butyloxycarbonyl-piperidin-4-yl)methyl-3-chloro-5-fluorobenzamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-(tert-Butyloxycarbonyl-piperidin-4-yl)methyl-3-chloro-5-fluorobenzamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.