accessibility menu, dialog, popup

UserName

CAS RN 18146-00-4

IUPAC Name: trimethyl(prop-2-en-1-yloxy)silane
Synonyms: trimethyl(prop-2-en-1-yloxy)silane
Molecular Weight (g/mol): 130.26
Molecular Formula: C6H14OSi
InChi Key: MNMVKGDEKPPREK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OCC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results