accessibility menu, dialog, popup

UserName

CH3CH3H3CSiCH3CH3HO

CAS RN 18173-64-3

IUPAC Name: tert-butyldimethylsilanol
Synonyms: tert-butyldimethylsilanol
Molecular Weight (g/mol): 132.28
Molecular Formula: C6H16OSi
InChi Key: FGWRMMTYIZKYMA-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)