accessibility menu, dialog, popup

UserName

NHBrBrF

CAS RN 1858251-75-8

IUPAC Name: 3,5-dibromo-N-[(4-fluorophenyl)methyl]aniline
Synonyms: 3,5-dibromo-N-[(4-fluorophenyl)methyl]aniline
Molecular Weight (g/mol): 359.04
Molecular Formula: C13H10Br2FN
InChi Key: VBVKQLQBFBSBIU-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC2=CC(Br)=CC(Br)=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results