accessibility menu, dialog, popup

UserName

CH3SOOCl

CAS RN 186551-47-3

IUPAC Name: 3'-methyl-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms: 3'-methyl-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol): 266.74
Molecular Formula: C13H11ClO2S
InChi Key: OHVJVSGIYVAZFD-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)