missing translation for 'onlineSavingsMsg'
Learn More

3'-Methylbiphenyl-4-sulfonyl chloride, 96%, Thermo Scientific™

Product Code. 15487327
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487327 250 mg 250mg
15467327 1 g 1g
15477327 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15487327 Supplier Thermo Scientific Alfa Aesar Supplier No. H34394.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186551-47-3
Molecular Formula C13H11ClO2S
Molecular Weight (g/mol) 266.739
MDL Number MFCD06409269
InChI Key OHVJVSGIYVAZFD-UHFFFAOYSA-N
Synonym 3'-methyl-biphenyl-4-sulfonyl chloride, 3'-methylbiphenyl-4-sulfonyl chloride, 3'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methyl, 4-3-methylphenyl benzenesulfonyl chloride, 3/'-methylbiphenyl-4-sulfonyl chloride, 3'-methyl 1,1'-biphenyl-4-sulfonyl chloride
PubChem CID 10754271
IUPAC Name 4-(3-methylphenyl)benzenesulfonyl chloride
SMILES CC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Melting Point 69°C to 73°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 266.74
Percent Purity 96%
Chemical Name or Material 3'-Methylbiphenyl-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.