accessibility menu, dialog, popup

UserName

ONHSOONFFFCl

CAS RN 188591-46-0

IUPAC Name: 4-chloro-N-(2-{[5-(trifluoromethyl)pyridin-2-yl]sulfonyl}ethyl)benzamide
Synonyms: 4-chloro-N-{2-[5-(trifluoromethyl)pyridin-2-ylsulfonyl]ethyl}benzamide
Molecular Weight (g/mol): 392.78
Molecular Formula: C15H12ClF3N2O3S
InChi Key: JFUIMTGOQCQTPF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=C1)S(=O)(=O)CCNC(=O)C1=CC=C(Cl)C=C1
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results