accessibility menu, dialog, popup

UserName

CAS RN 18883-66-4

CH3NNOOHN(R)(S)OHO(R)OH(S)OH(R)HO

CAS RN 18883-66-4

IUPAC Name: 3-methyl-3-nitroso-1-[(2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea
Synonyms: streptozocin streptozocin preparation streptozocin (substance) zanosar streptozotocine streptozocin (TN zanosar) pharmacia brand of streptozocin STZ SZC SZN
Molecular Weight (g/mol): 265.22
Molecular Formula: C8H15N3O7
InChi Key: ZSJLQEPLLKMAKR-GKHCUFPYSA-N
SMILES: CN(N=O)C(=O)N[C@H]1[C@@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O
Color 
  • (6)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (6)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)