CAS RN 188923-01-5
CAS RN 188923-01-5
IUPAC Name:
({5-[(2-{1-[3,5-bis(trifluoromethyl)phenyl]ethoxy}-3-(4-fluorophenyl)morpholin-4-yl)methyl]-2H-1,2,3-triazol-4-yl}methyl)dimethylamine hydrochloride
Synonyms:
({5-[(2-{1-[3,5-bis(trifluoromethyl)phenyl]ethoxy}-3-(4-fluorophenyl)morpholin-4-yl)methyl]-2H-1,2,3-triazol-4-yl}methyl)dimethylamine hydrochloride
Molecular Weight (g/mol):
611.99
Molecular Formula:
C26H29ClF7N5O2
InChi Key:
VZBKOBSSEVXFNF-UHFFFAOYNA-N
SMILES:
Cl.CC(OC1OCCN(CC2=NNN=C2CN(C)C)C1C1=CC=C(F)C=C1)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F