missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L 760735

Product Code. 18085932
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18085932

Brand: Tocris Bioscience™ 4013/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity NK1 antagonist

High affinity NK1 receptor antagonist (IC50 = 0.19 nM at human NK1 receptors). Selective (>300-fold) over h-NK2 and h-NK3 receptors. Exhibits anxiolytic and antidepressant-like effects. Orally active.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188923-01-5
Molecular Formula C26H29ClF7N5O2

Specifications

Quantity 10 mg
Formula Weight 611.98
Purity >99%
Molecular Weight (g/mol) 611.99
Chemical Name or Material L 760735
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.