accessibility menu, dialog, popup

UserName

CAS RN 189940-24-7

CH3OOCH3H3CNHOO

CAS RN 189940-24-7

IUPAC Name: 5-(4-methoxy-3-propoxyphenyl)-5-methyl-1,3-oxazolidin-2-one
Synonyms: 5-(4-methoxy-3-propoxyphenyl)-5-methyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: PCCPERGCFKIYIS-UHFFFAOYNA-N
SMILES: CCCOC1=CC(=CC=C1OC)C1(C)CNC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results