missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Mesopram

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Orally active phosphodiesterase (PDE) 4 inhibitor. Inhibits Th1 cell proliferation and decreases production of IFN-γ, TNF-α, IL-10 and iNOS in vitro. Triggers ovulation and exhibits efficacy against experimental autoimmune encephalomyelitis and murine colitis in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 265.31 |
| Purity | >98% |
| Molecular Weight (g/mol) | 265.309 |
| Chemical Name or Material | Mesopram |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction