CAS RN 1910124-24-1
CAS RN 1910124-24-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ GSK 6853
Potent and selective BRPF1 inhibitor
2
GSK6853, MedChemExpress
MedChemExpress GSK6853 is a potent and selective inhibitor of the BRPF1 bromodomain.