CAS RN 192-51-8
CAS RN 192-51-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dibenzo[e,l]pyrene, TRC
CAS: 192-51-8 Molecular Formula: C24 H14 Molecular Weight (g/mol): 302.368 Synonym: Dibenzo[fg,op]naphthacene,Dibenzo(e,l)pyrene,Dibenzo[fg,op]tetracene; SMILES: c1ccc2c(c1)c3cccc4c5ccccc5c6cccc2c6c34