missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[e,l]pyrene, TRC

Product Code. 30136205
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136205 50 mg 50mg
17959164 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136205 Supplier Toronto Research Chemicals Supplier No. TRCD41737550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192-51-8
Molecular Formula C24 H14
Molecular Weight (g/mol) 302.368
Synonym Dibenzo[fg,op]naphthacene, Dibenzo(e,l)pyrene, Dibenzo[fg,op]tetracene;
SMILES c1ccc2c(c1)c3cccc4c5ccccc5c6cccc2c6c34

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H14/c1-2-8-16-15(7-1)19-11-5-13-21-17-9-3-4-10-18(17)22-14-6-12-20(16)24(22)23(19)21/h1-14H
Formula Weight 302.11
Chemical Name or Material Dibenzo[e,l]pyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.