accessibility menu, dialog, popup

UserName

CAS RN 1940-18-7

CH3HO

CAS RN 1940-18-7

IUPAC Name: 1-ethylcyclohexan-1-ol
Synonyms: 1-ethylcyclohexanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: BUCJHJXFXUZJHL-UHFFFAOYSA-N
SMILES: CCC1(O)CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results