accessibility menu, dialog, popup

UserName

H3CCH3OHCH3CH3

CAS RN 19549-72-5

IUPAC Name: 2,4-dimethylheptan-3-ol
Synonyms: 2,4-dimethylheptan-3-ol
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: PSCLFHNWSAXNJL-UHFFFAOYNA-N
SMILES: CCCC(C)C(O)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results