accessibility menu, dialog, popup

UserName

OSN

CAS RN 19989-67-4

IUPAC Name: 1,3-benzothiazole-6-carbaldehyde
Synonyms: 1,3-benzothiazole-6-carbaldehyde
Molecular Weight (g/mol): 163.19
Molecular Formula: C8H5NOS
InChi Key: AVSFPLJXSHRMHM-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2N=CSC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results