accessibility menu, dialog, popup

UserName

NOOHOOH(E/Z)

CAS RN 201216-39-9

IUPAC Name: 4-phenyl-1-(4-phenylbutyl)piperidine; but-2-enedioic acid
Synonyms: 4-ppbp; butenedioic acid
Molecular Weight (g/mol): 409.53
Molecular Formula: C25H31NO4
InChi Key: OASPNIMFGJVLES-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(O)=O.C(CCC1=CC=CC=C1)CN1CCC(CC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results