accessibility menu, dialog, popup

UserName

BOHNFFFK

CAS RN 2017555-07-4

IUPAC Name: potassium [4-(benzylcarbamoyl)phenyl]trifluoroboranuide
Synonyms: potassium [4-(benzylcarbamoyl)phenyl]trifluoroboranuide
Molecular Weight (g/mol): 317.16
Molecular Formula: C14H12BF3KNO
InChi Key: RDEFKKXMPRJFFH-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results