CAS RN 202409-31-2
CAS RN 202409-31-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Desmethyl Etoricoxib, TRC
CAS: 202409-31-2 Molecular Formula: C17 H13 Cl N2 O2 S Molecular Weight (g/mol): 344.82 Synonym: 5-Chloro-3-[4-(methylsulfonyl)phenyl]-2,3'-bipyridine,5-Chloro-3-(4-methylsulfonylphenyl)-2-(3-pyridyl)pyridine,6'-Desmethyletoricoxib,Desmethyl Etoricoxib IUPAC Name: 5-chloro-3-(4-methylsulfonylphenyl)-2-pyridin-3-ylpyridine SMILES: CS(=O)(=O)c1ccc(cc1)c2cc(Cl)cnc2c3cccnc3
2
5-Chloro-3-(4-methylsulfonylphenyl)-2-(pyridin-3-yl)pyridine (6’-Desmethyletoricoxib), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.