missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Etoricoxib, TRC

Product Code. 30132607
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132607 50 mg 50mg
30006778 10 mg 10mg
30011662 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132607 Supplier Toronto Research Chemicals Supplier No. TRCD29689550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202409-31-2
Molecular Formula C17 H13 Cl N2 O2 S
Molecular Weight (g/mol) 344.82
Synonym 5-Chloro-3-[4-(methylsulfonyl)phenyl]-2,3'-bipyridine, 5-Chloro-3-(4-methylsulfonylphenyl)-2-(3-pyridyl)pyridine, 6'-Desmethyletoricoxib, Desmethyl Etoricoxib
IUPAC Name 5-chloro-3-(4-methylsulfonylphenyl)-2-pyridin-3-ylpyridine
SMILES CS(=O)(=O)c1ccc(cc1)c2cc(Cl)cnc2c3cccnc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H13ClN2O2S/c1-23(21,22)15-6-4-12(5-7-15)16-9-14(18)11-20-17(16)13-3-2-8-19-10-13/h2-11H,1H3
Formula Weight 344.0386
Percent Purity >95%
Chemical Name or Material 6'-Desmethyletoricoxib
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.