CAS RN 2039-87-4
CAS RN 2039-87-4
IUPAC Name:
1-chloro-2-ethenylbenzene
Synonyms:
O-chlorostyrene
Molecular Weight (g/mol):
138.59
Molecular Formula:
C8H7Cl
InChi Key:
ISRGONDNXBCDBM-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC=C1C=C
1
–
1
of
1
results
1
2-Chlorostyrene, 95%, stab. with 0.1% 4-tert-butylcatechol
CAS: 2039-87-4 Molecular Formula: C8H7Cl Molecular Weight (g/mol): 138.594 MDL Number: MFCD00000567 InChI Key: ISRGONDNXBCDBM-UHFFFAOYSA-N Synonym: 2-chlorostyrene,1-chloro-2-vinylbenzene,o-chlorostyrene,benzene, 1-chloro-2-ethenyl,styrene, o-chloro,ortho-chlorostyrene,stryene, o-chloro,styrene, 2-chloro,unii-za43r4q315,ccris 4598 PubChem CID: 14906 IUPAC Name: 1-chloro-2-ethenylbenzene SMILES: C=CC1=CC=CC=C1Cl