Learn More
2-Chlorostyrene, 95%, stab. with 0.1% 4-tert-butylcatechol
Product Code. 11447134
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11447134
Brand: Thermo Scientific Alfa Aesar B21551.14
Request Bulk or Custom Format
CAS: 2039-87-4 | C8H7Cl | 138.594 g/mol
2-Chlorostyrene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chlorostyrene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2039-87-4
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.594
MDL Number MFCD00000567
InChI Key ISRGONDNXBCDBM-UHFFFAOYSA-N
Synonym 2-chlorostyrene, 1-chloro-2-vinylbenzene, o-chlorostyrene, benzene, 1-chloro-2-ethenyl, styrene, o-chloro, ortho-chlorostyrene, stryene, o-chloro, styrene, 2-chloro, unii-za43r4q315, ccris 4598
PubChem CID 14906
IUPAC Name 1-chloro-2-ethenylbenzene
SMILES C=CC1=CC=CC=C1Cl
Specifications
Melting Point -63°C
Density 1.091
Boiling Point 189°C
Flash Point 65°C (149°F)
Odor Pungent
Refractive Index 1.565
Quantity 25 g
Beilstein 2038863
Solubility Information Insoluble in water.
Formula Weight 138.6
Percent Purity 98%
Chemical Name or Material 2-Chlorostyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorostyrene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : o-Chlorostyrene